Gene Transcripts

Gene Symbol
RNF44
Entrez Gene ID
22838
Gene Name
ring finger protein 44
Gene Aliases
Chromosome Location
Chr. 5: 176526699 - 176543456 on Build GRCh38
UniGene ID
Hs.434888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014901.4 NP_055716.1 11 2844
XM_005265840.2 XP_005265897.1 11 2556
XM_005265841.4 XP_005265898.1 11 2801
XM_005265842.4 XP_005265899.1 11 2786
XM_005265843.2 XP_005265900.1 11 2492
XM_005265844.3 XP_005265901.1 11 2597
XM_005265845.4 XP_005265902.1 11 2735
XM_006714831.2 XP_006714894.1 11 2547
XM_006714832.2 XP_006714895.1 11 2415
XM_011534466.2 XP_011532768.1 11 2559
XM_011534467.2 XP_011532769.1 11 2790
GenBank mRNA AB029023.1 - 11 2729
AK127673.1 - 2 1666
BC039833.1 - 11 2800 5535911
BC063297.1 - 11 2845 30349418

View Full Product Details