Gene Transcripts

Gene Symbol
TOB2
Entrez Gene ID
10766
Gene Name
transducer of ERBB2, 2
Gene Aliases
APRO5, TOB4, TOBL, TROB2
Chromosome Location
Chr. 22: 41433488 - 41448230 on Build GRCh38
UniGene ID
Hs.474978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016272.3 NP_057356.1 2 2234
XM_005261315.2 XP_005261372.1 2 3445
XM_006724105.3 XP_006724168.1 2 2751
XM_017028539.1 XP_016884028.1 2 4527
GenBank mRNA AB051450.1 - 2 1993
BC038957.1 - 2 2007 5786640

View Full Product Details