Gene Transcripts
- Gene Symbol
- BAIAP3
- Entrez Gene ID
- 8938
- Gene Name
- BAI1 associated protein 3
- Gene Aliases
- BAP3
- Chromosome Location
- Chr. 16: 1333605 - 1349441 on Build GRCh38
- UniGene ID
- Hs.458427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199096.1 |
NP_001186025.1
|
5, 6 |
616 |
|
NM_001199097.1 |
NP_001186026.1
|
5, 6 |
558 |
|
NM_001199098.1 |
NP_001186027.1
|
5, 6 |
489 |
|
NM_001199099.1 |
NP_001186028.1
|
5, 6 |
474 |
|
NM_001286464.1 |
NP_001273393.1
|
5, 6 |
609 |
|
NM_003933.4 |
NP_003924.2
|
5, 6 |
666 |
|
XM_011522728.1 |
XP_011521030.1
|
5, 6 |
1212 |
|
XM_011522729.1 |
XP_011521031.1
|
5, 6 |
1212 |
|
XM_011522730.2 |
XP_011521032.1
|
5, 6 |
1212 |
|
GenBank mRNA |
AB018277.2 |
- |
5, 6 |
518 |
|
AK294491.1 |
- |
5, 6 |
717 |
|
AK294744.1 |
- |
5, 6 |
434 |
|
AK295645.1 |
- |
5, 6 |
448 |
|
AK299309.1 |
- |
5, 6 |
518 |
|
AK299358.1 |
- |
5, 6 |
616 |
|
AK315059.1 |
- |
5, 6 |
666 |
|
AL834321.1 |
- |
5, 6 |
560 |
|
BC104966.1 |
- |
5, 6 |
529 |
8143969 |
BC112129.1 |
- |
5, 6 |
541 |
8327597 |
View Full Product Details