Gene Transcripts

Gene Symbol
BCL7C
Entrez Gene ID
9274
Gene Name
BCL tumor suppressor 7C
Gene Aliases
Chromosome Location
Chr. 16: 30834041 - 30894550 on Build GRCh38
UniGene ID
Hs.658547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004765.3 NP_004756.2 6 904
XM_017023885.1 XP_016879374.1 6 1155
GenBank mRNA AJ223980.1 - 6 546
BC019071.1 - 6 680 4909589

View Full Product Details