Gene Transcripts

Gene Symbol
GPC6
Entrez Gene ID
10082
Gene Name
glypican 6
Gene Aliases
OMIMD1
Chromosome Location
Chr. 13: 93226825 - 94408020 on Build GRCh38
UniGene ID
Hs.444329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005708.3 NP_005699.1 7 1841
XM_011521044.2 XP_011519346.1 7 1166
XM_017020298.1 XP_016875787.1 7 3449
XM_017020299.1 XP_016875788.1 7 2216
XM_017020300.1 XP_016875789.1 7 1073
XM_017020301.1 XP_016875790.1 6 1044
XM_017020302.1 XP_016875791.1 4 838
GenBank mRNA AF105267.1 - 7 1794
AF111178.1 - 7 1843
AK074741.1 - 8 1295
AK290144.1 - 7 1362
AY358462.1 - 7 1774
BC106947.2 - 7 1689 40034564

View Full Product Details