Gene Transcripts
- Gene Symbol
- ARPC2
- Entrez Gene ID
- 10109
- Gene Name
- actin related protein 2/3 complex subunit 2
- Gene Aliases
- ARC34, PNAS-139, PRO2446, p34-Arc
- Chromosome Location
- Chr. 2: 218217094 - 218254348 on Build GRCh38
- UniGene ID
- Hs.529303
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005731.3 |
NP_005722.1
|
5, 6 |
534 |
|
NM_152862.2 |
NP_690601.1
|
6, 7 |
629 |
|
XM_011510471.1 |
XP_011508773.1
|
2, 3 |
811 |
|
XM_017003113.1 |
XP_016858602.1
|
5, 6 |
503 |
|
XM_017003114.1 |
XP_016858603.1
|
6, 7 |
2912 |
|
XM_017003115.1 |
XP_016858604.1
|
5, 6 |
493 |
|
GenBank mRNA |
AF006085.1 |
- |
5, 6 |
544 |
|
AF279893.1 |
- |
4, 5 |
386 |
|
AK092520.1 |
- |
2, 3 |
1591 |
|
AK093625.1 |
- |
2, 3 |
1582 |
|
AK290268.1 |
- |
6, 7 |
528 |
|
BC000590.2 |
- |
6, 7 |
492 |
3346685 |
BT006898.1 |
- |
5, 6 |
460 |
|
U50523.1 |
- |
6, 7 |
544 |
|
View Full Product Details