Gene Transcripts
- Gene Symbol
- ATP6AP2
- Entrez Gene ID
- 10159
- Gene Name
- ATPase H+ transporting accessory protein 2
- Gene Aliases
- APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
- Chromosome Location
- Chr. X: 40580964 - 40606637 on Build GRCh38
- UniGene ID
- Hs.495960
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005765.2 |
NP_005756.2
|
8, 9 |
956 |
|
GenBank mRNA |
AF109363.1 |
- |
8, 9 |
918 |
|
AF291814.1 |
- |
8, 9 |
943 |
|
AK075382.1 |
- |
8, 9 |
1014 |
|
AK293510.1 |
- |
6, 7 |
704 |
|
AK296614.1 |
- |
7, 8 |
819 |
|
AK297007.1 |
- |
9, 10 |
815 |
|
AK297760.1 |
- |
7, 8 |
930 |
|
AK314633.1 |
- |
9, 10 |
990 |
|
AK315948.1 |
- |
7, 8 |
791 |
|
AY038990.1 |
- |
8, 9 |
959 |
|
AY429341.1 |
- |
7, 8 |
819 |
|
BC084541.1 |
- |
8, 9 |
936 |
6575732 |
Y17975.1 |
- |
3, 4 |
295 |
|
View Full Product Details