Gene Transcripts
- Gene Symbol
- DOCK4
- Entrez Gene ID
- 9732
- Gene Name
- dedicator of cytokinesis 4
- Gene Aliases
- Chromosome Location
- Chr. 7: 111726108 - 112206407 on Build GRCh38
- UniGene ID
- Hs.654652
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014705.3 |
NP_055520.3
|
7, 8 |
814 |
|
XM_006716188.2 |
XP_006716251.1
|
7, 8 |
801 |
|
XM_006716189.2 |
XP_006716252.1
|
7, 8 |
801 |
|
XM_017012819.1 |
XP_016868308.1
|
7, 8 |
1174 |
|
XM_017012820.1 |
XP_016868309.1
|
7, 8 |
737 |
|
XM_017012821.1 |
XP_016868310.1
|
7, 8 |
728 |
|
XM_017012822.1 |
XP_016868311.1
|
7, 8 |
1319 |
|
XM_017012823.1 |
XP_016868312.1
|
7, 8 |
728 |
|
XM_017012824.1 |
XP_016868313.1
|
7, 8 |
728 |
|
XM_017012825.1 |
XP_016868314.1
|
7, 8 |
728 |
|
GenBank mRNA |
AK309605.1 |
- |
7, 8 |
814 |
|
AY233380.1 |
- |
7, 8 |
545 |
|
BC117688.2 |
- |
7, 8 |
545 |
40072000 |
BC117689.1 |
- |
7, 8 |
545 |
40072006 |
BC117694.1 |
- |
7, 8 |
545 |
40072219 |
View Full Product Details