Gene Transcripts
- Gene Symbol
- BMS1
- Entrez Gene ID
- 9790
- Gene Name
- BMS1, ribosome biogenesis factor
- Gene Aliases
- ACC, BMS1L
- Chromosome Location
- Chr. 10: 42782506 - 42834937 on Build GRCh38
- UniGene ID
- Hs.10848
- Species
- Homo sapiens
- Gene Symbol
- BMS1P5
- Entrez Gene ID
- 399761
- Gene Name
- BMS1, ribosome biogenesis factor pseudogene 5
- Gene Aliases
- BMS1LP1, BMS1LP5, BMS1P1, bA556L1.3
- Chromosome Location
- Chr. 10: 46786733 - 46811989 on Build GRCh38
- UniGene ID
- Hs.711898
- Species
- Homo sapiens
- Gene Symbol
- BMS1P6
- Entrez Gene ID
- 642826
- Gene Name
- BMS1, ribosome biogenesis factor pseudogene 6
- Gene Aliases
- BMS1LP2, BMS1LP6, BMS1P2, bA302K17.2
- Chromosome Location
- Chr. 10: 47535574 - 47553523 on Build GRCh38
- UniGene ID
- Hs.463017
- Species
- Homo sapiens
View Full Product Details