Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005751.4 | NP_005742.4 | 2 | 364 | |
NM_147185.2 | NP_671714.1 | 2 | 364 | ||
XM_017011642.1 | XP_016867131.1 | 2 | 372 | ||
XM_017011643.1 | XP_016867132.1 | 2 | 372 | ||
XM_017011644.1 | XP_016867133.1 | 2 | 372 | ||
XM_017011645.1 | XP_016867134.1 | 2 | 372 | ||
XM_017011646.1 | XP_016867135.1 | 2 | 372 | ||
XM_017011647.1 | XP_016867136.1 | 2 | 372 | ||
XM_017011648.1 | XP_016867137.1 | 2 | 372 | ||
XM_017011649.1 | XP_016867138.1 | 2 | 372 | ||
XM_017011650.1 | XP_016867139.1 | 2 | 372 | ||
XM_017011651.1 | XP_016867140.1 | 2 | 372 | ||
XM_017011652.1 | XP_016867141.1 | 2 | 372 | ||
XM_017011653.1 | XP_016867142.1 | 2 | 372 | ||
XM_017011654.1 | XP_016867143.1 | 2 | 372 | ||
GenBank mRNA | AB019691.1 | - | 2 | 225 | |
AF026245.1 | - | 3 | 359 | ||
AF247727.4 | - | 2 | 139 | ||
AJ131693.1 | - | 2 | 361 | ||
BC015533.2 | - | 2 | 313 | ||
CR997756.1 | - | 2 | 364 |