Gene Transcripts
- Gene Symbol
- THBS4
- Entrez Gene ID
- 7060
- Gene Name
- thrombospondin 4
- Gene Aliases
- TSP-4, TSP4
- Chromosome Location
- Chr. 5: 79991296 - 80083287 on Build GRCh38
- UniGene ID
- Hs.211426
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306212.1 |
NP_001293141.1
|
15, 16 |
1907 |
|
NM_001306213.1 |
NP_001293142.1
|
15, 16 |
1959 |
|
NM_001306214.1 |
NP_001293143.1
|
14, 15 |
1901 |
|
NM_003248.5 |
NP_003239.2
|
14, 15 |
2073 |
|
XM_017009798.1 |
XP_016865287.1
|
14, 15 |
2053 |
|
XM_017009799.1 |
XP_016865288.1
|
14, 15 |
2053 |
|
XM_017009800.1 |
XP_016865289.1
|
14, 15 |
2053 |
|
GenBank mRNA |
AK302842.1 |
- |
14, 15 |
1901 |
|
AK313587.1 |
- |
14, 15 |
1889 |
|
BC050456.1 |
- |
15, 16 |
2157 |
6191808 |
Z19585.1 |
- |
14, 15 |
1861 |
|
View Full Product Details