Gene Transcripts

Gene Symbol
AKAP4
Entrez Gene ID
8852
Gene Name
A-kinase anchoring protein 4
Gene Aliases
AKAP 82, AKAP-4, AKAP82, CT99, FSC1, HI, PRKA4, hAKAP82, p82
Chromosome Location
Chr. X: 50190768 - 50201013 on Build GRCh38
UniGene ID
Hs.97633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003886.2 NP_003877.2 2 229
NM_139289.1 NP_647450.1 2 202
GenBank mRNA AF072756.1 - 2 257
AK292463.1 - 2 223
BC033007.1 - 2 226
BC070150.1 - 2 221
BC126250.1 - 2 228 8991966
BC126252.1 - 2 228 8991968
Y15195.1 - 2 229

View Full Product Details