Gene Transcripts

Gene Symbol
VSNL1
Entrez Gene ID
7447
Gene Name
visinin like 1
Gene Aliases
HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1
Chromosome Location
Chr. 2: 17540540 - 17656439 on Build GRCh38
UniGene ID
Hs.444212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003385.4 NP_003376.2 4 1919
GenBank mRNA BC022012.2 - 4 1760
BF055598.1 - 98

View Full Product Details