Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001326578.1 | NP_001313507.1 | 10 | 1210 | |
NM_001326579.1 | NP_001313508.1 | 10 | 997 | ||
NM_003569.2 | NP_003560.2 | 10 | 1133 | ||
NR_137169.1 | 9 | 1060 | |||
GenBank mRNA | AK301464.1 | - | 4 | 496 | |
BC011975.1 | - | 10 | 997 | 3860059 | |
U77942.1 | - | 10 | 962 |