Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195303.2 | NP_001182232.1 | 3 | 3022 | |
NM_001254748.1 | NP_001241677.1 | 6 | 3080 | ||
NM_001254749.1 | NP_001241678.1 | 5 | 3207 | ||
NM_003617.3 | NP_003608.1 | 5 | 3195 | ||
GenBank mRNA | BU187973.1 | - | 550 | ||
BX537427.1 | - | 5 | 2939 |