Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277226.1 | NP_001264155.1 | 11 | 1333 | |
NM_001277227.1 | NP_001264156.1 | 11 | 1189 | ||
NM_003667.3 | NP_003658.1 | 12 | 1405 | ||
NR_110596.1 | 13 | 1689 | |||
GenBank mRNA | AB105190.1 | - | 12 | 1374 | |
AF061444.1 | - | 12 | 1094 | ||
AF062006.1 | - | 12 | 1142 | ||
AK075399.1 | - | 13 | 1475 | ||
AK294985.1 | - | 12 | 1406 | ||
AK315133.1 | - | 12 | 1382 | ||
BC096324.3 | - | 12 | 1140 | 40008253 | |
BC096325.1 | - | 11 | 1068 | 40008255 | |
BC096326.3 | - | 11 | 1068 | 40008257 | |
BC099650.1 | - | 12 | 1140 | 40008256 | |
FN820440.1 | - | 11 | 926 |