Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008910.2 | NP_001008910.1 | 8 | 1352 | |
NR_026909.1 | 8 | 1617 | |||
XM_011512067.2 | XP_011510369.1 | 7 | 1257 | ||
XM_017005160.1 | XP_016860649.1 | 7 | 1452 | ||
GenBank mRNA | AF060798.1 | - | 8 | 1323 | |
AF203910.1 | - | 8 | 1319 | ||
AJ005791.1 | - | 7 | 1310 | ||
AJ010872.1 | - | 7 | 1225 | ||
AK096954.1 | - | 6 | 2026 | ||
AK225392.1 | - | 8 | 1317 | ||
AK225526.1 | - | 1157 | |||
AK292694.1 | - | 8 | 1352 | ||
BC002618.2 | - | 8 | 1321 | 3143105 | |
BC053998.1 | - | 8 | 1317 | 5760866 | |
CN268939.1 | - | 1 | 46 |