Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001272095.1 | NP_001259024.1 | 7 | 1092 | |
NM_001272096.1 | NP_001259025.1 | 9 | 978 | ||
NM_004604.4 | NP_004595.2 | 8 | 956 | ||
XM_005255521.1 | XP_005255578.1 | 8 | 873 | ||
XM_011545925.2 | XP_011544227.1 | 4 | 382 | ||
GenBank mRNA | AF026007.1 | - | 8 | 641 | |
AF318489.1 | - | 7 | 1015 | ||
AJ000541.1 | - | 7 | 777 | ||
AK091833.1 | - | 9 | 978 | ||
AK296652.1 | - | 7 | 1012 | ||
AK315716.1 | - | 8 | 911 | ||
BC002436.2 | - | 8 | 865 | 3345863 | |
BT007326.1 | - | 8 | 641 | ||
CR541806.1 | - | 8 | 641 | ||
U07158.1 | - | 8 | 707 | ||
X85784.1 | - | 8 | 641 |