Gene Transcripts

Gene Symbol
TSPAN7
Entrez Gene ID
7102
Gene Name
tetraspanin 7
Gene Aliases
A15, CCG-B7, CD231, DXS1692E, MRX58, MXS1, TALLA-1, TM4SF2, TM4SF2b
Chromosome Location
Chr. X: 38561478 - 38688918 on Build GRCh38
UniGene ID
Hs.441664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004615.3 NP_004606.2 5, 6 674
GenBank mRNA AB062057.1 - 5, 6 675
AB102665.1 - 5, 6 590
AK293178.1 - 6, 7 659
AK293534.1 - 7, 8 828
AK293552.1 - 6, 7 739
AK312343.1 - 5, 6 608
BC018036.1 - 5, 6 633 4801388
D10653.1 - 5, 6 614
D29808.1 - 5, 6 600
L10373.1 - 5, 6 658

View Full Product Details