Gene Transcripts

Gene Symbol
SRPX
Entrez Gene ID
8406
Gene Name
sushi repeat containing protein, X-linked
Gene Aliases
DRS, ETX1, HEL-S-83p, SRPX1
Chromosome Location
Chr. X: 38149335 - 38220924 on Build GRCh38
UniGene ID
Hs.15154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170750.1 NP_001164221.1 2 402
NM_001170751.1 NP_001164222.1 3 462
NM_001170752.1 NP_001164223.1 3 462
NM_006307.4 NP_006298.1 3 462
XM_017029893.1 XP_016885382.1 3 401
GenBank mRNA AK125542.1 - 2 383
AK289430.1 - 3 409
AK293127.1 - 3 439
AK298802.1 - 3 398
BC020684.1 - 3 417 4731946
DA422854.1 - 2 375
DC388035.1 - 3 462
EU668359.1 - 3 368
U61374.1 - 3 368
U78093.1 - 3 369

View Full Product Details