Gene Transcripts

Gene Symbol
ZNF75A
Entrez Gene ID
7627
Gene Name
zinc finger protein 75a
Gene Aliases
Chromosome Location
Chr. 16: 3305406 - 3322999 on Build GRCh38
UniGene ID
Hs.513292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302109.1 NP_001289038.1 7 2607
NM_001324041.1 NP_001310970.1 7 2564
NM_001324085.1 NP_001311014.1 5 2300
NM_153028.3 NP_694573.1 6 2083
NR_126069.1 5 2242
NR_136681.1 6 2411
NR_136683.1 5 1887
NR_136686.1 6 2437
XM_011522646.2 XP_011520948.1 6 2496
XM_011522647.2 XP_011520949.1 6 2181
XM_017023642.1 XP_016879131.1 6 1979
GenBank mRNA AK056091.1 - 6 2006
AK093979.1 - 5 2195
CR749782.1 - 6 1906

View Full Product Details