Gene Transcripts

Gene Symbol
C17orf80
Entrez Gene ID
55028
Gene Name
chromosome 17 open reading frame 80
Gene Aliases
HLC-8, MIG3, SPEP1
Chromosome Location
Chr. 17: 73232233 - 73248959 on Build GRCh38
UniGene ID
Hs.12929
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100622.2 NP_001094092.1 4 1745
NM_017941.5 NP_060411.2 4 1745
NR_110105.1 3 225
XM_005257487.3 XP_005257544.1 4 1738
XM_011524961.1 XP_011523263.1 4 1745
XM_011524962.2 XP_011523264.1 3 1643
XM_017024806.1 XP_016880295.1 3 1671
GenBank mRNA AK000728.1 - 4 1704
AK027166.1 - 2 339
AY239293.1 - 2 415
BC005005.2 - 3 1650 3544728
BG754800.1 - 2 142

View Full Product Details