Gene Transcripts

Gene Symbol
SMG8
Entrez Gene ID
55181
Gene Name
SMG8, nonsense mediated mRNA decay factor
Gene Aliases
C17orf71
Chromosome Location
Chr. 17: 59210010 - 59215250 on Build GRCh38
UniGene ID
Hs.7296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018149.6 NP_060619.4 3 2421
GenBank mRNA AF349467.1 - 3 2421
AK001449.1 - 3 1570
AK026858.1 - 3 2394
AL834490.1 - 3 2400
BC020957.1 - 3 1589 3851898

View Full Product Details