Gene Transcripts
- Gene Symbol
- PRR11
- Entrez Gene ID
- 55771
- Gene Name
- proline rich 11
- Gene Aliases
- Chromosome Location
- Chr. 17: 59155499 - 59206709 on Build GRCh38
- UniGene ID
- Hs.631750
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018304.3 |
NP_060774.2
|
10 |
3687 |
|
GenBank mRNA |
AK001891.1 |
- |
11 |
1553 |
|
View Full Product Details