Gene Transcripts
- Gene Symbol
- RRN3
- Entrez Gene ID
- 54700
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor
- Gene Aliases
- A-270G1.2, TIFIA
- Chromosome Location
- Chr. 16: 15060022 - 15094335 on Build GRCh38
- UniGene ID
- Hs.460078
- Species
- Homo sapiens
- Gene Symbol
- RRN3P1
- Entrez Gene ID
- 730092
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 21796630 - 21819174 on Build GRCh38
- UniGene ID
- Hs.31290
- Species
- Homo sapiens
- Gene Symbol
- RRN3P2
- Entrez Gene ID
- 653390
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 16: 29074842 - 29116718 on Build GRCh38
- UniGene ID
- Hs.720348
- Species
- Homo sapiens
View Full Product Details