Gene Transcripts

Gene Symbol
ARMC7
Entrez Gene ID
79637
Gene Name
armadillo repeat containing 7
Gene Aliases
Chromosome Location
Chr. 17: 75109952 - 75130265 on Build GRCh38
UniGene ID
Hs.413047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304271.1 NP_001291200.1 2093
NM_024585.3 NP_078861.1 2157
GenBank mRNA BC011728.1 - 2122

View Full Product Details