Gene Transcripts

Gene Symbol
ATAD5
Entrez Gene ID
79915
Gene Name
ATPase family, AAA domain containing 5
Gene Aliases
C17orf41, ELG1, FRAG1
Chromosome Location
Chr. 17: 30830932 - 30896424 on Build GRCh38
UniGene ID
Hs.528902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024857.4 NP_079133.3 2 1331
XM_011525269.2 XP_011523571.1 2 1352
XM_011525270.2 XP_011523572.1 2 1352
XM_011525273.2 XP_011523575.1 2 2369
XM_017025111.1 XP_016880600.1 2 1327
GenBank mRNA AJ314648.2 - 2 1299
AL832103.1 - 2 1332
AY557611.1 - 2 1299
BC015051.1 - 2 1320

View Full Product Details