Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142448.1 | NP_001135920.1 | 4 | 510 | |
NM_001142449.1 | NP_001135921.1 | 2 | 681 | ||
NM_001142450.1 | NP_001135922.1 | 4 | 660 | ||
NM_001142451.1 | NP_001135923.1 | 3 | 747 | ||
NM_032038.2 | NP_114427.1 | 3 | 747 | ||
XM_006721096.3 | XP_006721159.1 | 4 | 517 | ||
XM_017023757.1 | XP_016879246.1 | 4 | 518 | ||
XM_017023758.1 | XP_016879247.1 | 4 | 514 | ||
XM_017023759.1 | XP_016879248.1 | 4 | 513 | ||
GenBank mRNA | AF212371.1 | - | 3 | 694 | |
AF370423.1 | - | 4 | 642 | ||
AK095677.1 | - | 3 | 744 | ||
AK289787.1 | - | 3 | 742 | ||
AL390215.1 | - | 2 | 643 | ||
AL524908.3 | - | 4 | 635 | ||
BC006156.1 | - | 3 | 358 | ||
BC008325.1 | - | 3 | 572 | 3501826 | |
BC038961.1 | - | 4 | 524 | 6053095 | |
BC047741.1 | - | 3 | 721 | 5757339 | |
BC065235.1 | - | 4 | 492 | 6161778 |