Gene Transcripts

Gene Symbol
RNFT2
Entrez Gene ID
84900
Gene Name
ring finger protein, transmembrane 2
Gene Aliases
TMEM118
Chromosome Location
Chr. 12: 116738291 - 116853631 on Build GRCh38
UniGene ID
Hs.437195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109903.1 NP_001103373.1 7, 8 1123
NM_032814.3 NP_116203.2 7, 8 1123
GenBank mRNA AK027533.1 - 8, 9 1159
AK093164.1 - 3, 4 924

View Full Product Details