Gene Transcripts

Gene Symbol
ZCRB1
Entrez Gene ID
85437
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Gene Aliases
MADP-1, MADP1, RBM36, SNRNP31, ZCCHC19
Chromosome Location
Chr. 12: 42312086 - 42326130 on Build GRCh38
UniGene ID
Hs.496279
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033114.3 NP_149105.3 8 1558
GenBank mRNA BC022543.1 - 8 1533

View Full Product Details