Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109763.1 | NP_001103233.1 | 5 | 846 | |
NM_001323900.1 | NP_001310829.1 | 5 | 846 | ||
NM_001323901.1 | NP_001310830.1 | 4 | 693 | ||
NM_144675.2 | NP_653276.1 | 4 | 462 | ||
XM_011545750.2 | XP_011544052.1 | 5 | 840 | ||
GenBank mRNA | AK128775.1 | - | 5 | 467 | |
AY302134.1 | - | 3 | 296 | ||
AY358206.1 | - | 4 | 685 | ||
BC016460.1 | - | 4 | 462 | 4150450 | |
BI752847.1 | - | 6 | 669 | ||
CV024949.1 | - | 3 | 296 |