Gene Transcripts

Gene Symbol
RILPL2
Entrez Gene ID
196383
Gene Name
Rab interacting lysosomal protein like 2
Gene Aliases
RLP2
Chromosome Location
Chr. 12: 123408567 - 123436733 on Build GRCh38
UniGene ID
Hs.488173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145058.2 NP_659495.1 4 1148
NR_130703.1 3 996
GenBank mRNA AB085763.1 - 4 911
AK056934.1 - 4 1132
BC013042.2 - 4 921 3544761

View Full Product Details