Gene Transcripts
- Gene Symbol
- LRRC57
- Entrez Gene ID
- 255252
- Gene Name
- leucine rich repeat containing 57
- Gene Aliases
- Chromosome Location
- Chr. 15: 42528332 - 42548804 on Build GRCh38
- UniGene ID
- Hs.234681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153260.2 |
NP_694992.2
|
4, 5 |
619 |
|
XM_011521423.2 |
XP_011519725.1
|
3, 4 |
507 |
|
XM_011521424.2 |
XP_011519726.1
|
3, 4 |
507 |
|
GenBank mRNA |
AK094131.1 |
- |
4, 5 |
613 |
|
AK094891.1 |
- |
3, 4 |
853 |
|
AK309861.1 |
- |
3, 4 |
1184 |
|
BC058935.1 |
- |
4, 5 |
575 |
5578024 |
BX538324.1 |
- |
3, 4 |
765 |
|
CN350663.1 |
- |
1, 2 |
83 |
|
CR749223.1 |
- |
4, 5 |
588 |
|
View Full Product Details