Gene Transcripts

Gene Symbol
TUSC5
Entrez Gene ID
286753
Gene Name
tumor suppressor candidate 5
Gene Aliases
DSPB1, IFITMD3, LOST1
Chromosome Location
Chr. 17: 1279663 - 1300987 on Build GRCh38
UniGene ID
Hs.146059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172367.2 NP_758955.2 3 1091
GenBank mRNA AB090231.1 - 3 1091
AL831929.1 - 3 575
BC142615.1 - 3 1075 40147346

View Full Product Details