Gene Transcripts

Gene Symbol
LINC01465
Entrez Gene ID
283416
Gene Name
long intergenic non-protein coding RNA 1465
Gene Aliases
C12orf61
Chromosome Location
Chr. 12: 62601751 - 62603434 on Build GRCh38
UniGene ID
Hs.375881
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_121682.1 1 470
GenBank mRNA AK098456.1 - 1 471
BC038738.1 - 1 173
BC136288.1 - 1 464 9020275

View Full Product Details