Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_194318.3 | NP_919299.3 | 12 | 1145 | |
XM_006719768.3 | XP_006719831.1 | 13 | 1335 | ||
XM_011534936.1 | XP_011533238.1 | 12 | 1134 | ||
XM_011534938.2 | XP_011533240.1 | 11 | 983 | ||
XM_017020395.1 | XP_016875884.1 | 13 | 1238 | ||
GenBank mRNA | AB101481.1 | - | 12 | 1035 | |
AK291273.1 | - | 12 | 1134 | ||
AY190526.1 | - | 12 | 1035 | ||
BC047395.1 | - | 10 | 1099 | ||
BC068595.1 | - | 12 | 1125 | 4837250 | |
DA290408.1 | - | 5 | 442 |