Gene Transcripts

Gene Symbol
CCDC57
Entrez Gene ID
284001
Gene Name
coiled-coil domain containing 57
Gene Aliases
Chromosome Location
Chr. 17: 82101458 - 82212892 on Build GRCh38
UniGene ID
Hs.631724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316321.1 NP_001303250.1 15 2893
GenBank mRNA AK074059.1 - 7 1350
BC040264.1 - 15 2877 5165045

View Full Product Details