Gene Transcripts

Gene Symbol
RPL22
Entrez Gene ID
6146
Gene Name
ribosomal protein L22
Gene Aliases
EAP, HBP15, HBP15/L22, L22
Chromosome Location
Chr. 1: 6185020 - 6199619 on Build GRCh38
UniGene ID
Hs.515329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000983.3 NP_000974.1 4 662
GenBank mRNA AF113701.1 - 1 125
AK097075.1 - 4 621
AL110153.1 - 1 62
BC035566.1 - 4 662 5109124
BC058887.1 - 4 620 6141451
BC066314.1 - 4 638 5303658

View Full Product Details