Gene Transcripts

Gene Symbol
PTPRD
Entrez Gene ID
5789
Gene Name
protein tyrosine phosphatase, receptor type D
Gene Aliases
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location
Chr. 9: 8314246 - 10612840 on Build GRCh38
UniGene ID
Hs.446083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040712.2 NP_001035802.1 2 261
NM_001171025.1 NP_001164496.1 2 261
NM_002839.3 NP_002830.1 13 901
NM_130391.3 NP_569075.2 2 261
NM_130392.3 NP_569076.2 2 261
NM_130393.3 NP_569077.2 2 261
XM_006716817.3 XP_006716880.1 12 1199
XM_006716823.2 XP_006716886.1 12 1193
XM_006716825.3 XP_006716888.1 12 1195
XM_006716827.3 XP_006716890.1 12 1192
XM_006716832.3 XP_006716895.1 12 1187
XM_006716833.3 XP_006716896.1 12 1189
XM_006716834.3 XP_006716897.1 12 1190
XM_006716835.3 XP_006716898.1 12 1189
XM_006716837.3 XP_006716900.1 12 1187
XM_006716838.3 XP_006716901.1 12 1189
XM_006716839.3 XP_006716902.1 12 1187
XM_011517992.2 XP_011516294.1 12 1192
XM_017014958.1 XP_016870447.1 12 1198
XM_017014959.1 XP_016870448.1 13 1270
XM_017014960.1 XP_016870449.1 11 1159
XM_017014961.1 XP_016870450.1 11 1143
XM_017014962.1 XP_016870451.1 3 331
XM_017014963.1 XP_016870452.1 12 1198
XM_017014964.1 XP_016870453.1 12 1197
XM_017014965.1 XP_016870454.1 12 1200
XM_017014966.1 XP_016870455.1 12 1198
XM_017014967.1 XP_016870456.1 12 1196
XM_017014968.1 XP_016870457.1 12 1195
XM_017014969.1 XP_016870458.1 12 1198
XM_017014970.1 XP_016870459.1 12 1198
XM_017014971.1 XP_016870460.1 12 1196
XM_017014972.1 XP_016870461.1 12 1198
XM_017014973.1 XP_016870462.1 12 1195
XM_017014974.1 XP_016870463.1 12 1198
XM_017014975.1 XP_016870464.1 12 1193
XM_017014976.1 XP_016870465.1 12 1195
XM_017014977.1 XP_016870466.1 12 1196
XM_017014978.1 XP_016870467.1 12 1195
XM_017014979.1 XP_016870468.1 12 1197
XM_017014980.1 XP_016870469.1 12 1192
XM_017014981.1 XP_016870470.1 12 1187
XM_017014982.1 XP_016870471.1 12 1194
XM_017014983.1 XP_016870472.1 12 1194
XM_017014984.1 XP_016870473.1 12 1194
XM_017014985.1 XP_016870474.1 12 1191
XM_017014986.1 XP_016870475.1 12 1190
XM_017014987.1 XP_016870476.1 12 1190
XM_017014988.1 XP_016870477.1 12 1190
XM_017014989.1 XP_016870478.1 12 1190
XM_017014990.1 XP_016870479.1 12 1189
XM_017014991.1 XP_016870480.1 12 1189
XM_017014992.1 XP_016870481.1 12 1189
XM_017014993.1 XP_016870482.1 12 1187
XM_017014994.1 XP_016870483.1 12 1187
XM_017014995.1 XP_016870484.1 12 1193
GenBank mRNA AB208869.1 - 1 87
AB211400.1 - 12 869
BC028038.1 - 973
BC106713.1 - 2 209 40027581
BC106714.1 - 2 209 40027582
BC106715.2 - 2 209 40027584
BC106716.1 - 2 209 40027585
DR001889.1 - 4 336
L38929.1 - 3 311

View Full Product Details