Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039936.2 | NP_001035025.1 | 7 | 1734 | |
NM_001293069.1 | NP_001279998.1 | 14 | 2005 | ||
NM_001293070.1 | NP_001279999.1 | 14 | 2219 | ||
NM_001293071.1 | NP_001280000.1 | 12 | 2124 | ||
NM_001293072.1 | NP_001280001.1 | 13 | 1937 | ||
NM_001293073.1 | NP_001280002.1 | 7 | 1599 | ||
NM_001293075.1 | NP_001280004.1 | 6 | 1461 | ||
NM_001293076.1 | NP_001280005.1 | 6 | 1560 | ||
NM_001293077.1 | NP_001280006.1 | 5 | 1518 | ||
NM_001293078.1 | NP_001280007.1 | 5 | 1422 | ||
NM_001293079.1 | NP_001280008.1 | 5 | 1490 | ||
NM_001293080.1 | NP_001280009.1 | 6 | 1596 | ||
NM_001293081.1 | NP_001280010.1 | 7 | 1249 | ||
NM_004067.3 | NP_004058.1 | 13 | 2180 | ||
NR_120524.1 | 4 | 1270 | |||
NR_120525.1 | 5 | 1340 | |||
XM_011515105.2 | XP_011513407.1 | 16 | 2060 | ||
XM_011515106.2 | XP_011513408.1 | 15 | 2021 | ||
XM_011515107.2 | XP_011513409.1 | 15 | 2803 | ||
XM_017011721.1 | XP_016867210.1 | 17 | 2078 | ||
XM_017011722.1 | XP_016867211.1 | 16 | 2821 | ||
GenBank mRNA | AK026415.1 | - | 7 | 1734 | |
AK126784.1 | - | 2 | 2938 | ||
AK294216.1 | - | 7 | 1249 | ||
AK294271.1 | - | 14 | 2006 | ||
AK316030.1 | - | 7 | 1258 | ||
U28926.1 | - | 13 | 1972 |