Gene Transcripts

Gene Symbol
SH3BP2
Entrez Gene ID
6452
Gene Name
SH3 domain binding protein 2
Gene Aliases
3BP-2, 3BP2, CRBM, CRPM, RES4-23
Chromosome Location
Chr. 4: 2793023 - 2841096 on Build GRCh38
UniGene ID
Hs.167679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122681.1 NP_001116153.1 12, 13 1676
NM_001145855.1 NP_001139327.1 12, 13 1819
NM_001145856.1 NP_001139328.1 12, 13 1766
NM_003023.4 NP_003014.3 12, 13 1817
GenBank mRNA AB000462.1 - 12, 13 1817
AB000463.1 - 13, 14 1996
AF000936.1 - 11, 12 1556
AK124073.1 - 7, 8 1662
AK225497.1 - 12, 13 1678
AK299996.1 - 12, 13 1735
AK312286.1 - 12, 13 1614
BC022996.1 - 12, 13 1672 5103986
U56386.1 - 12, 13 1764

View Full Product Details