Gene Transcripts

Gene Symbol
RTN2
Entrez Gene ID
6253
Gene Name
reticulon 2
Gene Aliases
NSP2, NSPL1, NSPLI, SPG12
Chromosome Location
Chr. 19: 45485288 - 45497055 on Build GRCh38
UniGene ID
Hs.47517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005619.4 NP_005610.1 10, 11 1790
NM_206900.2 NP_996783.1 9, 10 1571
NM_206901.2 NP_996784.1 6, 7 613
GenBank mRNA AF004222.1 - 10, 11 1689
AF004223.1 - 7, 8 839
AF004224.1 - 6, 7 581
AF038540.1 - 7, 8 1020
AK291967.1 - 9, 10 1552
AK314158.1 - 9, 10 1554
BC073874.1 - 10, 11 1732 5242827

View Full Product Details