Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000422.2 | NP_000413.1 | 8 | 1350 | |
GenBank mRNA | AK095342.1 | - | 8 | 1279 | |
BC000159.2 | - | 8 | 1279 | 2986100 | |
BC011901.2 | - | 8 | 1277 | 4111703 | |
BC056421.1 | - | 8 | 1279 | 5208304 | |
BC072018.1 | - | 9 | 1217 | 3531173 | |
BC072019.1 | - | 8 | 1275 | 5442690 | |
BX647923.1 | - | 7 | 1616 | ||
X05803.1 | - | 7 | 731 | ||
X62571.1 | - | 8 | 1277 |