Gene Transcripts

Gene Symbol
PGR
Entrez Gene ID
5241
Gene Name
progesterone receptor
Gene Aliases
NR3C3, PR
Chromosome Location
Chr. 11: 101029624 - 101130681 on Build GRCh38
UniGene ID
Hs.32405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000926.4 NP_000917.3 7 3285
NM_001202474.3 NP_001189403.1 7 2535
NM_001271161.2 NP_001258090.1 6 2229
NM_001271162.1 NP_001258091.1 7 1205
NR_073141.2 7 2483
NR_073142.2 6 2366
NR_073143.2 5 2098
XM_006718858.3 XP_006718921.1 6 4004
GenBank mRNA AB084248.1 - 6 2236
AB085683.1 - 5 2105
AB085843.1 - 6 2373
AB085844.1 - 6 2132
AB085845.1 - 7 2490
AF016381.1 - 7 2717
AK304853.1 - 7 1205
AY212933.1 - 4 1915
M15716.1 - 7 2717
X51730.1 - 7 3996

View Full Product Details