Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002419.3 | NP_002410.1 | 10 | 3175 | |
GenBank mRNA | AK057256.1 | - | 5 | 1415 | |
AK090614.1 | - | 9 | 1744 | ||
AK222781.1 | - | 10 | 3168 | ||
AK299609.1 | - | 10 | 2204 | ||
AK316032.1 | - | 10 | 2044 | ||
BC011263.2 | - | 10 | 3163 | 4215281 | |
BC064543.1 | - | 10 | 3134 | 5733866 | |
L32976.1 | - | 10 | 3163 | ||
U07747.1 | - | 10 | 3132 |