Gene Transcripts

Gene Symbol
NPAT
Entrez Gene ID
4863
Gene Name
nuclear protein, coactivator of histone transcription
Gene Aliases
E14, E14/NPAT, p220
Chromosome Location
Chr. 11: 108155280 - 108222638 on Build GRCh38
UniGene ID
Hs.171061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321307.1 NP_001308236.1 6 543
NM_002519.2 NP_002510.2 6 543
XM_011542854.2 XP_011541156.1 6 543
XM_017017802.1 XP_016873291.1 6 1281
XM_017017803.1 XP_016873292.1 6 724
GenBank mRNA AK290020.1 - 6 543
AK291687.1 - 6 467
BC040356.1 - 6 534
BC050561.2 - 6 528
BC136270.1 - 6 464 9020257
D83243.1 - 6 507
DA338219.1 - 6 543
U58852.1 - 6 441
X97186.1 - 6 475

View Full Product Details