Gene Transcripts
- Gene Symbol
- MYO9A
- Entrez Gene ID
- 4649
- Gene Name
- myosin IXA
- Gene Aliases
- Chromosome Location
- Chr. 15: 71822290 - 72118577 on Build GRCh38
- UniGene ID
- Hs.513000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006901.3 |
NP_008832.2
|
4, 5 |
1496 |
|
XM_006720539.2 |
XP_006720602.1
|
4, 5 |
1496 |
|
XM_011521613.2 |
XP_011519915.1
|
4, 5 |
1496 |
|
XM_011521614.2 |
XP_011519916.1
|
5, 6 |
1182 |
|
XM_011521615.2 |
XP_011519917.1
|
4, 5 |
1349 |
|
XM_011521616.2 |
XP_011519918.1
|
4, 5 |
1143 |
|
XM_011521617.2 |
XP_011519919.1
|
4, 5 |
1496 |
|
XM_011521618.2 |
XP_011519920.1
|
4, 5 |
1496 |
|
XM_011521619.2 |
XP_011519921.1
|
4, 5 |
1496 |
|
XM_011521620.2 |
XP_011519922.1
|
4, 5 |
1496 |
|
XM_011521621.2 |
XP_011519923.1
|
4, 5 |
1496 |
|
XM_011521622.2 |
XP_011519924.1
|
4, 5 |
1496 |
|
XM_017022228.1 |
XP_016877717.1
|
5, 6 |
1182 |
|
XM_017022229.1 |
XP_016877718.1
|
5, 6 |
1331 |
|
XM_017022230.1 |
XP_016877719.1
|
4, 5 |
1496 |
|
GenBank mRNA |
AB209841.1 |
- |
3, 4 |
931 |
|
AB290183.1 |
- |
3, 4 |
931 |
|
AF117888.1 |
- |
4, 5 |
1247 |
|
AJ001714.1 |
- |
3, 4 |
1112 |
|
BC060886.1 |
- |
4, 5 |
1462 |
|
BC140869.1 |
- |
4, 5 |
1165 |
9021737 |
BC150294.1 |
- |
3, 4 |
486 |
|
DQ088983.1 |
- |
2, 3 |
133 |
|
DQ088984.1 |
- |
2, 3 |
133 |
|
View Full Product Details