Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001294345.1 | NP_001281274.1 | 2 | 215 | |
NM_001294346.1 | NP_001281275.1 | 2 | 322 | ||
NM_016533.5 | NP_057617.2 | 2 | 561 | ||
GenBank mRNA | AK296012.1 | - | 2 | 561 | |
AK298043.1 | - | 2 | 273 | ||
AK315995.1 | - | 2 | 559 | ||
BC057766.1 | - | 2 | 229 | ||
BI597990.1 | - | 2 | 414 | ||
BM128205.1 | - | 2 | 183 | ||
CB995632.1 | - | 2 | 242 | ||
CD695373.1 | - | 2 | 309 |