Gene Transcripts

Gene Symbol
FZD2
Entrez Gene ID
2535
Gene Name
frizzled class receptor 2
Gene Aliases
Fz2, fz-2, fzE2, hFz2
Chromosome Location
Chr. 17: 44557444 - 44561262 on Build GRCh38
UniGene ID
Hs.142912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001466.3 NP_001457.1 1 1047
GenBank mRNA AB017364.1 - 1 840
BC052266.1 - 1 1032
BC113402.1 - 2 849 8322454
BC113404.1 - 2 849 8322456
L37882.1 - 1 934

View Full Product Details