Gene Transcripts

Gene Symbol
HSPG2
Entrez Gene ID
3339
Gene Name
heparan sulfate proteoglycan 2
Gene Aliases
HSPG, PLC, PRCAN, SJA, SJS, SJS1
Chromosome Location
Chr. 1: 21822232 - 21937297 on Build GRCh38
UniGene ID
Hs.562227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291860.1 NP_001278789.1 76 10475
NM_005529.6 NP_005520.4 76 10472
XM_011541318.2 XP_011539620.1 81 10956
XM_017001120.1 XP_016856609.1 77 10602
XM_017001121.1 XP_016856610.1 76 10551
XM_017001122.1 XP_016856611.1 76 10548
GenBank mRNA AB209851.1 - 28 4212
M85289.1 - 76 10472
X62515.1 - 76 10438

View Full Product Details